missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18501-1-AP-20UL
This item is not returnable.
View return policy
Description
HOXA9 is the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development.Specifications
| HOXA9 | |
| Polyclonal | |
| Unconjugated | |
| HOXA9 | |
| ABD B, homeobox A9, Homeobox protein Hox 1G, Homeobox protein Hox A9, HOX1, HOX1.7, HOX1G, HOXA9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15405, 297099, 3205 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P09631, P31269 | |
| HOXA9 | |
| HOXA9 Fusion Protein Ag13350 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction