missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXA9 Monoclonal Antibody (1A12C1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531766
This item is not returnable.
View return policy
Description
MA5-31766 has been tested in indirect ELISA.
HOXA9 is the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development.
Specifications
| HOXA9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P31269 | |
| Hoxa9 | |
| Purified recombinant fragment of human HOXA9 (AA: 1-272) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 1A12C1 | |
| Unconjugated | |
| Hoxa9 | |
| ABD-B; D6a9; homeo box A9; homeobox A9; homeobox A9-like; homeobox protein Hox-1.7; Homeobox protein Hox-1G; homeobox protein Hox-A9; homeodomain protein HOXA9; HOX1; HOX1.7; Hox-1.7; HOX1G; HOXA 9; Hoxa9; Hoxa-9; Hoxa-9 protein; Hoxa9l; RGD1310001 | |
| Mouse | |
| Protein G | |
| RUO | |
| 3205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction