missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101101
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HOXA7.
Hox genes play a fundamental role in the development of the vertebrate central nervous system, heart, axial skeleton, limbs, gut, urogenital tract and external genitalia. HoxA2 protein may contribute to dorsal-ventral patterning and/or to the specification of neuronal phenotype. HoxA7 functions as a potent transcriptional repressor and its action as such requires several domains, including both activator and repressor regions. HoxA7 is expressed in the fetal liver, lung, skeletal muscle, kidney, pancreas and placenta.
Specifications
| HOXA7 | |
| Polyclonal | |
| Unconjugated | |
| HOXA7 | |
| ANTP; AV118143; homeo box A7; homeobox A7; homeobox A9; Homeobox gene A7; homeobox protein Hox 1.1; Homeobox protein Hox-1.1; homeobox protein HOX-1A; Homeobox protein Hox-A7; HOMEOBOX PROTEIN HOX-A7 (HOX-1.1) (M6-12) (M6); Homeobox protein M6-12; homeobox protein R5; HOX1; HOX1.1; hox-1.1; HOX1A; Hox1r5; Hoxa7; Hoxa-7; Hoxa9; M6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15404, 3204, 500126 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P02830, P09634, P31268 | |
| HOXA7 | |
| A synthesized peptide derived from human HOXA7(Accession P31268), corresponding to amino acid residues G107-R157. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction