missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA7 Mouse anti-Human, CoraLite 488, Clone: 2A7H6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67112-100UL
This item is not returnable.
View return policy
Description
HOXA7 Monoclonal antibody apecifically detetects HOXA7 antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| HOXA7 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| HOXA7 | |
| HOXA7 fusion protein Ag19025 | |
| 100 μL | |
| 3204 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Immunofluorescence | |
| 2A7H6 | |
| CoraLite 488 | |
| BC148692 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction