missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18210-1-AP-150UL
This item is not returnable.
View return policy
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.Specifications
| HOXA6 | |
| Polyclonal | |
| Unconjugated | |
| HOXA6 | |
| homeobox A6, Homeobox protein Hox 1B, Homeobox protein Hox A6, HOX1, HOX1.2, HOX1B, HOXA6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15403, 3203 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09092, P31267 | |
| HOXA6 | |
| HOXA6 Fusion Protein Ag12875 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction