missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXA6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101639
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HOXA6.
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
Specifications
| HOXA6 | |
| Polyclonal | |
| Unconjugated | |
| HOXA6 | |
| homeo box 1B; homeo box A6; homeobox A6; homeobox protein Hox-1.2; Homeobox protein Hox-1B; homeobox protein HOXA6; homeobox protein Hox-A6; homeobox protein M5-4; HOX1; HOX1.2; Hox-1.2; HOX1B; Hoxa6; Hoxa-6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3203 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31267 | |
| HOXA6 | |
| A synthesized peptide derived from human HOXA6(Accession P31267), corresponding to amino acid residues L103-S153. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction