missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA5 Polyclonal Antibody

HOXA5 Polyclonal Antibody
Brand: Invitrogen PA5101638
This item is not returnable.
View return policy
Description
HOXA5 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| HOXA5 | |
| Polyclonal | |
| Unconjugated | |
| HOXA5 | |
| homeo box 1C; homeo box A5; homeobox A5; homeobox protein Hox-1.3; Homeobox protein Hox-1C; homeobox protein HOXA5; homeobox protein Hox-A5; homeobox protein M2; HOX1; HOX1.3; Hox-1.3; HOX1C; Hox-1C; Hoxa5; Hoxa-5; Unknown (protein for MGC:138007) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15402, 3202, 79241 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09021, P20719, P52949 | |
| HOXA5 | |
| A synthesized peptide derived from human HOXA5(Accession P20719), corresponding to amino acid residues H63-D113. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction