missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£305.00 - £410.00
Specifications
| Antigen | HOXA5 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18477010
|
Novus Biologicals
NBP1-83235-25ul |
25 μL |
£305.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18777983
|
Novus Biologicals
NBP1-83235 |
£410.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
HOXA5 Polyclonal specifically detects HOXA5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| HOXA5 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| homeo box 1C, homeo box A5, homeobox A5, Homeobox protein Hox-1C, homeobox protein Hox-A5, homeobox protein HOXA5, HOX1, HOX1.3, HOX1CMGC9376 | |
| HOXA5 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 3202 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MHSGRYGYGYNGMDLSVGRSGSGHFGSGERARSYAASASAAPAEPRYSQPATSTHSPQPDPLPCSAVAPSPGSDSHHGGKNSLSNSSGASADAGSTHISSREGVGTASGAEEDAPASSEQASAQSEPSPAPPAQPQIYPWMRKLHISHDN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title