missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXA10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115375
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HOXA10.
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factorm HOXA10, that may regulate gene expression, morphogenesis, and differentiation. More specifically, HOXA10 may function in fertility, embryo viability, and regulation of hematopoietic lineage commitment. Alternatively spliced transcript variants have been described. Read-through transcription also exists between this gene and the downstream homeobox A9 gene.
Specifications
| HOXA10 | |
| Polyclonal | |
| Unconjugated | |
| HOXA10 | |
| homeo box A10; homeobox A10; homeobox protein 1H; homeobox protein Hox-1.8; Homeobox protein Hox-1H; homeobox protein HOXA10; homeobox protein Hox-A10; HOX1; HOX1.8; Hox-1.8; Hox-18; HOX1H; HOXA10; Hoxa-10; PL | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3206 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P31260 | |
| HOXA10 | |
| A synthesized peptide derived from human HOXA10(Accession P31260), corresponding to amino acid residues A263-S313. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction