missing translation for 'onlineSavingsMsg'
Learn More

HOXA10 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-06103PCP

Product Code. 18675916

  • £326.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HOXA10 Polyclonal antibody specifically detects HOXA10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HOXA10
Polyclonal
Immunohistochemistry-Paraffin
homeo box A10, homeobox A10, homeobox protein 1H, Homeobox protein Hox-1.8, Homeobox protein Hox-1H, homeobox protein Hox-A10, homeobox protein HOXA10, HOX1, HOX1HHOX1.8, MGC12859, PL
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human HOXA10.
0.1 mL
Primary
Human
Purified
Immunohistochemistry (Paraffin)
PerCP
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
3206
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.