missing translation for 'onlineSavingsMsg'
Learn More

HOX11 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18421887 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421887 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421887 Supplier Novus Biologicals Supplier No. NBP281887G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HOX11 Polyclonal antibody specifically detects HOX11 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOX11
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias homeo box 11 (T-cell lymphoma 3-associated breakpoint), homeo box-11 (T-cell leukemia-3 associated breakpoint, homologous to DrosophilaNotch), Homeobox protein Hox-11, HOX11T-cell leukemia, homeobox 1, Proto-oncogene TCL-3, T-cell leukemia homeobox 1, T-cell leukemia homeobox protein 1, T-cell leukemia/lymphoma protein 3, TCL3MGC163402
Host Species Rabbit
Immunogen HOX11 antibody was raised against a 16 amino acid peptide near the amino terminus of human HOX11. The immunogen is located within the first 50 amino acids of HOX11. Amino Acid Squence: MGPASRKQDGEYGLG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 3195
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.