missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Howard Leight Sync™ Electo-H Helmet Digital FM Radio and Stereo

Product Code. 15896425
Change view
Click to view available options
:
Black, SNR 29 dB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15896425 Black, SNR 29 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15896425 Supplier Honeywell™ Supplier No. 1030334
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Hearing protector with built-in stereo FM radio.

  • Combines FM radio earmuff with advanced sound amplification technology
  • Allows wearers to hear important communications other co-workers, alarms and important warning signals at a safely amplified level, while listening to the radio
  • Positioned microphones for natural, directional hearing
  • Advanced response technology amplifies incoming sounds to 82 dB, then acts as a passive earmuff for sounds above 85 dB
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Certifications/Compliance EC Category PPE: 1, EN 352
Noise Reduction Rating (NRR) SNR: 29 dB
Includes Requires 2 x AA batteries
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.