missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Howard Leight™ Leightning L3HV Hi-Visibility Headband Ear Muff
Description
- Exclusive padded foam headband and super soft ear cushions eliminate pressure on head
- Air Flow Control™ technology delivers optimal attenuation across all frequencies
- Robust steel-wire construction withstands demanding use
- Snap-in ear cushions replace easily
- Telescoping height adjustment offers personalized comfort
Specifications
Specifications
| Color | Fluorescent Green |
| For Use With (Application) | Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel |
| Noise Reduction Rating (NRR) | 30 dB |
| Product Type | Ear Muffs |
| Dielectric | No |
| Style | Over-the-head |
| Includes | Reflective padded PUR-E, PVC headband |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?