Learn More
Honeywell™ Howard Leight™ Leightning L1 Ear Muffs
Product Code. 12665083
Change view
Click to view available options
L1N; NRR 25dB
Product Code.
12665083 L1N; NRR 25dB
1 options
Product Code. 12665083 Supplier Honeywell™ Supplier No. 1011994
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High performance and robust steel wire durability. This noise-blocking earmuff meets NRR 25 Canada Class B requirements.
  • Robust steel wire construction withstands demanding use, especially in tough environments
  • Features patented Air Flow Control technology, delivering optimal attenuation across all frequencies
  • Snap-in ear cushions make replacement quick and easy
  • Multiple attenuation levels allow for targeted attenuation for a variety of applications
  • Telescopic height adjustment remains fixed during use
  • Delivers high degree of comfort; ideal for situations where compact earmuffs and reliable Noise-Blocking protection are required
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Certifications/Compliance ANSI S3.19-1974
For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel
Noise Reduction Rating (NRR) 25 dB
Product Type Ear Muffs
Dielectric No
Style Behind-the-neck
Includes Elastic headband strap