missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HotTaq DNA Polymerase
Click to view available options
Quantity:
1000 units
2500 units
500 units
Unit Size:
1000 units
2500 units
500 units
Description
- The enzyme has been proved not having the 3′ - 5′exonuclease activity.
- Prior the first PCR step the HotTaq DNA polymerase should be activated by 15 minute incubation at 95-97°C.
Specifications
Specifications
| Concentration | 10 U/μL |
| Content And Storage | Store at -20°C Guaranteed stable for 6 months when properly stored |
| Quantity | 1000 units |
| For Use With (Application) | Standard PCR |
| Storage Buffer | 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction