missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HotTaq DNA Polymerase

Product Code. 16002555
Change view
Click to view available options
Quantity:
1000 units
2500 units
500 units
Unit Size:
1000 units
2500 units
500 units
Product Code. Quantity unitSize
16002555 2500 units 2500 units
16012555 500 units 500 units
16092545 1000 units 1000 units
3 options
This item is not returnable. View return policy

Product Code. 16002555

Brand: Abnova™ D0011.2500Units

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HotTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.

  • The enzyme has been proved not having the 3′ - 5′exonuclease activity.
  • Prior the first PCR step the HotTaq DNA polymerase should be activated by 15 minute incubation at 95-97°C.

Specifications

Concentration 10 U/μL
Content And Storage Store at -20°C Guaranteed stable for 6 months when properly stored
Quantity 2500 units
For Use With (Application) Standard PCR
Storage Buffer 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.