missing translation for 'onlineSavingsMsg'
Learn More

Stuart™ Hotplate Accessories

Product Code. 15125914
Change view
Click to view available options
Size:
Small
5 mL
10 mL
25 mL
Type:
Block Base
Block Insert
Clamp, Heater Block
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Type unitSize
15125914 5 mL Block Insert Each
15105924 10 mL Block Insert Each
15135924 25 mL Block Insert Each
15135934 Small Clamp, Heater Block Each
15105914 Small Block Base Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15125914 Supplier Stuart™ Supplier No. 0480598
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Hotplate Accessories for use with Hotplates/Stirrers that have a 6 x 6 in. top plate.

  • This heating block system with interchangeable inserts can be used with various Stuart Hotplates and Hotplate Stirrers
  • Designed to heat round-bottom flasks
  • Small and medium sized base plates offered; each can accommodate up to three block inserts each
  • A third larger base plate features heat-resistant handles and accepts a single block insert for flasks up to 500 mL
  • Chemical- and solvent-resistant anodized block inserts provide a safe method for synthesis of single or multiple reactions
  • Hotplate accessories offer equal or better heating performance than silicon oil
  • The low-well provides excellent temperature transfer without compromising the view of the reaction or risk of cracking the glassware
  • Each base plate features a hole to house a temperature controller probe

Specifications

For Use With (Equipment) Hot Plates (6 x 6 in.)
Quantity 3 Pk.
Size 5 mL
Type Block Insert
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.