Learn More
Manutan Hot Gloves™, size nine
Product Code. 12846469
Encompass_Preferred
Product Code. 12846469 Supplier Manutan Supplier No. A351155
Only null left
Add to basket
Add to basket
Double layered cotton for extra protection and durability. Nitrile grip pattern offers good grip for safer handling. Ideal for bakeries, food manufacturing, oven applications, pressings, mouldings.
  • Contact heat-resistant to 250°C
  • EN 388, EN 407
  • Size 9