missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Host cell factor C2 Antibody (OTI5H9), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71862PE
This item is not returnable.
View return policy
Description
Host cell factor C2 Monoclonal antibody specifically detects Host cell factor C2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Host cell factor C2 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 29915 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI5H9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| FLJ94012, HCF2, HCF-2C2 factor, host cell factor 2, host cell factor C2 | |
| Full length human recombinant protein of human HCFC2(NP_037452) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction