missing translation for 'onlineSavingsMsg'
Learn More

Host cell factor C2 Antibody (OTI5H9), DyLight 594, Novus Biologicals™

Product Code. 18405327 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405327 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405327 Supplier Novus Biologicals Supplier No. NBP271862DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Host cell factor C2 Monoclonal antibody specifically detects Host cell factor C2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Host cell factor C2
Applications Western Blot, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone OTI5H9
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias FLJ94012, HCF2, HCF-2C2 factor, host cell factor 2, host cell factor C2
Host Species Mouse
Immunogen Full length recombinant protein of human Host cell factor C2 (NP_037452) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29915
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.