missing translation for 'onlineSavingsMsg'
Learn More

Host Cell Factor 1/HCFC1 Antibody [Biotin], Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals AF6254B

Product Code. 30615537

  • £482.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Host Cell Factor 1/HCFC1 Polyclonal antibody specifically detects Host Cell Factor 1/HCFC1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Host Cell Factor 1/HCFC1
Polyclonal
PBS
Goat
Antigen Affinity-purified
RUO
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunocytochemistry
Biotin
CFFC1 factor, HCF, HCF1HFC1, HCF-1VP16 accessory protein, host cell factor C1 (VP16-accessory protein), MGC70925, VCAFhost cell factor 1
E. coli-derived recombinant human Host Cell Factor 1/HCFC1, Ala1626-Val1836, Accession # P51610
0.1 mL
Cell Biology, Cell Cycle and Replication, Endocrinology
3054
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.