missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polypropylene Hose Nozzle for Buerkle™ Stopcock

Product Code. 10654765 Shop All Bürkle Products
Change view
Click to view available options
Diameter (Metric):
10 mm
16 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10654765 10 mm Each
10260272 16 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10654765 Supplier Buerkle™ Supplier No. 05251310
  • ONLINE EXCLUSIVE PRICE
    £15.55
    £14.73 / Pack of 1
    Save £0.82
    5% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Buerkle™ Blue Polypropylene Hose Nozzle is a water tight accessory for use with Buerkle™ Stopcock.

  • Interchangeable hose nozzles for StopCock taps
  • Secure, leak-proof connection via union nut (G 0.75 in.)
  • High flow rate, no constrictions other than the chosen nozzle diameter
TRUSTED_SUSTAINABILITY

Specifications

Type Hose Nozzle
Diameter (Metric) 10 mm
Description Blue
Material Polypropylene
For Use With (Equipment) Stop Cock
Connectors Hose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.