Learn More
Cole-Parmer Hose Barb Fittings, Straight Reducer, Stainless Steel
Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Outer:
1/8 in.
5/32 in.
Unit Size:
Pack of 10
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Quick and easy assembly without tools. Single barb design tightly grips the tubing. RoHS compliant
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS |
| Diameter (English) Inner | 3/32 in. |
| Diameter (English) Outer | 5/32 in. |
| Outlet Port | Hose Barb Reducer |
| Sterilization Method | E-beam/gamma irradiation, EtO, or autoclave up to 134°C |
| Shape | Straight |
| Color | Metallic |