missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Hose Adapter

Product Code. 10796114
Change view
Click to view available options
Color:
Gray
Green
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10796114 Green Each
10010461 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10796114 Supplier Hirschmann™ Supplier No. 9903103
  • ONLINE EXCLUSIVE PRICE
    £8.09
    £6.16 / Pack of 1
    Save £1.93
    23% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spare part for pipetus™ standard, hose connectors to pump

  • Spare part for pipetus™ standard, hose connectors to pump

Specifications

Color Green
For Use With (Equipment) Pipetus Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.