missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hornerin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£289.00 - £458.00
Specifications
| Antigen | Hornerin |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000, KnockDown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18408640
|
Novus Biologicals
NBP1-80807-25ul |
25ul |
£289.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18755603
|
Novus Biologicals
NBP1-80807 |
£458.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Hornerin Polyclonal specifically detects Hornerin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Hornerin | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| hornerin, intermediate filament-associated protein, S100A16, S100a18 | |
| HRNR | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000, KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q86YZ3 | |
| 388697 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:WSAGENDSYSRNVRGSLKPGTESISRRLSFQRDFSGQHNSYSGQSSSYGEQNSDSHQSSGRGQCGSGS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title