missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Horn shark IgNar Recombinant Rabbit Monoclonal Antibody (HL3138)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556852
This item is not returnable.
View return policy
Description
Horn shark IgNar Recombinant Monoclonal Antibody for Western Blot
Cartilaginous fish have evolved a distinctive type of immunoglobulin, designated as IgNAR, which lacks the light polypeptide chains and is composed entirely by heavy chains. IgNAR molecules can be manipulated by molecular engineering to produce the variable domain of a single heavy chain polypeptide (vNARs).
Specifications
| Horn shark IgNar | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Shark | |
| Antibody | |
| IgG |
| Western Blot | |
| HL3138 | |
| Unconjugated | |
| Horn shark IgNar; Nanobody; Single domain antibody | |
| Full length Horn shark IgNar recombinant protein(AAX10144.1). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction