missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Horizontal Mini-gel Unit

Product Code. 11795668 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric) Gel:
100 mm
160 mm
Width (Metric) Gel:
75 mm
140 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Gel Width (Metric) Gel unitSize
11795668 100 mm 75 mm Each
11719599 160 mm 140 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11795668 Supplier Cole-Parmer™ Supplier No. 2855910

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Type Horizontal Mini-gel Unit
Includes Safety cover with attached power leads, gel tray, tapered baffles for casting gels, and long-life platinum electrodes
Quantity 1 Each
Volume (Metric) Buffer 250 mL
Length (Metric) Gel 100 mm
Width (Metric) 98.5 mm
Width (Metric) Gel 75 mm
Height (Metric) 101.6 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.