missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOPX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11419-1-AP-150UL
This item is not returnable.
View return policy
Description
HOPX is an atypical homeodomain protein which does not bind DNA and is required to modulate cardiac growth and development. HOPX acts via its interaction with SRF, thereby modulating the expression of SRF-dependent cardiac-specific genes and cardiac development. HOPX prevents SRF-dependent transcription either by inhibiting SRF binding to DNA or by recruiting histone deacetylase (HDAC) proteins that prevent transcription by SRF. HOPX overexpression causes cardiac hypertrophy and may act as a tumor suppressor.Specifications
| HOPX | |
| Polyclonal | |
| Unconjugated | |
| HOPX | |
| CAMEO, HOD, Homeodomain only protein, HOP, HOP homeobox, HOPX, LAGY, NECC1, OB1, Odd homeobox protein 1, SMAP31, TOTO | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171160, 74318, 84525 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q78ZR5, Q8R1H0, Q9BPY8 | |
| Hopx | |
| HOPX Fusion Protein Ag1979 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction