missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hopx Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590538
This item is not returnable.
View return policy
Description
Immunogen sequence: MSAETASGPT EDQVEILEYN FNKVDKHPDS TTLCLIAAEA GLSEEETQKW FKQRLAKWRR SEGLPSECRS VTD; Positive Samples: mouse brain, mouse lung, rat lung; Cellular Location: Nucleus.
HOPX is an atypical homeodomain protein which does not bind DNA and is required to modulate cardiac growth and development. HOPX acts via its interaction with SRF, thereby modulating the expression of SRF-dependent cardiac-specific genes and cardiac development. HOPX prevents SRF-dependent transcription either by inhibiting SRF binding to DNA or by recruiting histone deacetylase (HDAC) proteins that prevent transcription by SRF. HOPX overexpression causes cardiac hypertrophy and may act as a tumor suppressor.
Specifications
| Hopx | |
| Polyclonal | |
| Unconjugated | |
| Hopx | |
| 1110018K11Rik; 1200015P04Rik; 2300002F06Rik; AI848177; AW490897; Cameo; GIIg15b; global ischemia induced protein GIIG15B; global ischemia-induced gene 15B protein; global ischemia-induced protein 15B; Hdop; HOD; homeobox only domain; homeobox-only protein; homeodomain only protein; homeodomain-only protein; Hop; HOP homeobox; Hopx; LAGY; Lung cancer-associated Y protein; mOB1; NECC1; not expressed in choriocarcinoma clone 1; Not expressed in choriocarcinoma protein 1; OB1; Obl; odd homeobox 1; odd homeobox protein 1; SMAP31; Toto | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 171160, 74318, 84525 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.61 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q78ZR5, Q8R1H0, Q9BPY8 | |
| Hopx | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction