missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ Hopen™ Pocketless Hygiene Gown
GREENER_CHOICE

Product Code. 16534051 Shop All Medicom Products
Click to view available options
Color:
Red
White
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 16534051

Brand: Medicom™ 40.301/03

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Medicom™ Hopen™ Pocketless Hygiene Gown available in three distinct colors

  • The Pocketless Hygiene gown protects the working environment from particles coming from the body of the operator
  • Allows the user to be protected from the risks of soiling related to the activity being carried out
  • Pleasant and light, polypropylene fabric is extremely breathable
  • Collared model without flap and elasticated at the wrists

Specifications

Size Large
Construction Made in China
For Use With (Application) Category 1 Personal Protective Equipment
Gender Unisex
No. of Pockets None
Packaging Type Individual, Bag
Product Type Hygiene Gown
Material Polypropylene
Color White
Closure Type Press Studs
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Seam Type Sewn
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.