missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ Hopen™ Double Elastic Bouffant cap

Product Code. 16504121 Shop All Medicom Products
Change view
Click to view available options
Color:
White
Yellow
Unit Size:
Pack of 1000
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16504121 Yellow Pack of 1000
17780129 White Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16504121 Supplier Medicom™ Supplier No. 70.104
  • ONLINE EXCLUSIVE PRICE
    £35.15
    £29.20 / Pack of 1000
    Save £5.95
    16% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medicom™ Hopen™ Double Elastic Bouffant cap available in four distinct colors

  • Lightweight, polypropylene bouffant cap provides additional comfort the user
  • Full head coverage ensures excellent retention for the user
  • Ability to use a color code for tracking
  • Can be used to control the environment for HACCP processes and various industries

Specifications

Product Type Clip Cap
Material Polypropylene
Color Yellow
Disposable Single-use
Seam Type Double Elastic
Sterility Non-sterile
For Use With (Equipment) Operating Room PPE
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Dimensions (L x W) 44 x 50 cm
Length (Metric) 44 cm
Width (Metric) 50 cm
Construction Made in China
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.