missing translation for 'onlineSavingsMsg'
Learn More

Hopantenate-d6 Calcium, TRC

Product Code. 30392507
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392507 5 mg 5mg
30400231 2.5 mg 2.5mg
30403253 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392507 Supplier Toronto Research Chemicals Supplier No. TRCH6695665MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hopantenate-d6 Calcium
Molecular Formula C20H24D12CaN2O10
Quantity 5 mg
Synonym 4-[[(2R)-2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl]amino]butanoic Acid Calcium Salt (2:1)-d6, 4-(2,4-Dihydroxy-3,3-dimethylbutyramido)butyric Acid Calcium Salt (2:1)-d6, Calcium D-(+)-Homopantothenate-d6, Calcium Homopantothenate-d6, Calcium Hopantenate-d6, D-(+)-Homopantothenic Acid Calcium Salt-d6, Pantocalcin-d6, Pantogam-d6, Vivant-d6
InChI Formula InChI=1S/2C10H19NO5.Ca/c2*1-10(2,6-12)8(15)9(16)11-5-3-4-7(13)14;/h2*8,12,15H,3-6H2,1-2H3,(H,11,16)(H,13,14);/q;;+2/p-2/t2*8-;/m00./s1/i2*3D2,4D2,5D2;
SMILES O=C([O-])C([2H])([2H])C([2H])([2H])C([2H])([2H])NC([C@H](O)C(C)(C)CO)=O.O=C([O-])C([2H])([2H])C([2H])([2H])C([2H])([2H])NC([C@H](O)C(C)(C)CO)=O.[Ca+2]
Molecular Weight (g/mol) 516.66
Formula Weight 516.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.