missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HOP Polyclonal antibody specifically detects HOP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | HOP |
| Applications | ELISA, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Gene Alias | CAMEO, HOD, homeodomain-only protein, HOP homeobox, HOPLAGYNECC1OB1SMAP31, Lung cancer-associated Y protein, MGC20820, not expressed in choriocarcinoma clone 1, Not expressed in choriocarcinoma protein 1, odd homeobox 1 protein, Odd homeobox protein 1, TOTO |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Gene ID (Entrez) | 84525 |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?