Learn More
Eisco™ Hooke's Law Apparatus
Product Code. 13162780
Change view
Click to view available options
15cm adjustable mirror on study 30cm support rod
Product Code.
13162780 15cm adjustable mirror on study 30cm support rod
1 options
Product Code. 13162780 Supplier Eisco™ Supplier No. PH0323
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Requires
Weights (not included)
Used to teach Hooke's Law in the classroom.
  • 15cm mirror scale is marked in millimeters and mounted on a sturdy 30 cm. support rod and wooden base
  • Mirror adjusts to prevent parallax error
  • Hook attached to rod supports a coiled spring with mass hanger and indicator
Specifications
Product Type Hooke's Law Spring Set
Includes 15 cm adjustable mirror on study 30 cm support rod