missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOOK3 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44279IR
This item is not returnable.
View return policy
Description
HOOK3 Polyclonal antibody specifically detects HOOK3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| HOOK3 | |
| Polyclonal | |
| Western Blot | |
| FLJ31058, hHK3, HK3h-hook3, hook homolog 3 (Drosophila), protein Hook homolog 3 | |
| The epitope recognized by this antibody maps to a region between residue 425 to 475 of human Protein Hook homolog 3 using the numbering given in entry Q86VS8.2 (GeneID 84376). | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| 84376 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction