missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOOK2 Recombinant Rabbit Monoclonal Antibody (23GB5965), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552958
This item is not returnable.
View return policy
Description
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
Specifications
| HOOK2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HOOK2 | |
| A630054I03Rik; hHK2; h-hook2; HK2; hook homolog 2; hook homolog 2 (Drosophila); hook microtubule tethering protein 2; hook microtubule-tethering protein 2; HOOK2; mHK2; protein Hook homolog 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170833, 29911, 304669 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5965 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7TMK6, Q96ED9 | |
| HOOK2 | |
| A synthesized peptide derived from human HOOK2 (35-62AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction