missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOOK1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10871-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the hook family of coiled-coil proteins, which bind to microtubules and organelles through their N- and C-terminal domains, respectively. The encoded protein localizes to discrete punctuate subcellular structures, and interacts with several members of the Rab GTPase family involved in endocytosis. It is thought to link endocytic membrane trafficking to the microtubule cytoskeleton. Several alternatively spliced transcript variants have been identified, but the full-length nature of some of these variants has not been determined.Specifications
| HOOK1 | |
| Polyclonal | |
| Unconjugated | |
| HOOK1 | |
| h hook1, hHK1, HK1, hook homolog 1 (Drosophila), HOOK1, Protein Hook homolog 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51361, 77963 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BIL5, Q9UJC3 | |
| HOOK1 | |
| HOOK1 Fusion Protein Ag1311 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction