missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ ThermoSafe™ Compact Insulated Shipper

Product Code. 11775509 Shop All Cole-Parmer Products
Click to view available options
Dimensions (L x W x H):
6 x 6 x 6in.
8 x 8 x 11in.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11775509

Brand: Cole-Parmer™ 0373030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Choose from a variety of sizes to meet your refrigeration needs with these compact insulated shippers. Cole Parmer™ ThermoSafe™ Compact Insulated Shippers are ideal for shipping non-hazardous materials.

  • Compact shippers have a cylindrical interior to hold tubes, vials and other small items
  • Add as many ice packs as you need
  • For non-hazardous materials only

Specifications

Height (English) 11 in.
Length (English) 8 in.
Width (English) 8 in.
Dimensions (L x W x H) 8 x 8 x 11in.
Diameter (English) 4.8 in.
Type Compact Insulated Shipper
Quantity 1 Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.