missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Temptation 505 SBEA Protective Footwear

Product Code. 15364392
Click to view available options
:
Size: 39
This item is not returnable. View return policy

Product Code. 15364392

Brand: Honeywell™ 655150539/7

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance EN ISO 20345:2011
Color Blue
Footbed Material Leather
For Use With (Application) For industrial use
Material Leather
Size 39
Sole Features Leather insole, excellent anti-skid properties
Sole Material Leather
Style Protective
Toe Material Steel
Resistant To Abrasion
Product Type Protective Footwear
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.