Learn More
Honeywell™ Pro'Clean S2 Shoes
Brand:  Honeywell™ 6246127-39/7
Code : NEW
Additional Details : Weight : 0.87300kg
 View more versions of this product
Product Code. 15269503
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
EN ISO 20345: 2011, SRC Certified
For professional use such as catering, industrial cleaning, laboratory, medical and pharmaceutical
39
Double density Polyurethane
Abrasion, fuel oil and grease
Shoes
White
Microfiber
Removable, anti-static insole
Spacium non-metallic
Yes