missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Pro'Clean S2 Shoes

Product Code. 15239503
Click to view available options
Size:
35
36
38
39
41
44
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 15239503

Brand: Honeywell™ 624612736/7

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance EN ISO 20345: 2011, SRC Certified
Color White
For Use With (Application) For professional use such as catering, industrial cleaning, laboratory, medical and pharmaceutical
Material Microfiber
Size 36
Sole Features Removable, anti-static insole
Sole Material Double density Polyurethane
Toe Material Spacium non-metallic
Resistant To Abrasion, fuel oil and grease
Waterproof Yes
Product Type Shoes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.