Learn More
Honeywell™ Freedom Half-Mask Air-Purifying Respirator (APR)
The Freedom Half-Mask APR is designed for occasional exposure to gases, vapours, and particulates. Limited use and maintenance-free, it is supplied pre-assembled with cartridges for elimination of assembly mistakes, replacement parts, and decontamination concerns.
Brand:  Honeywell™ 1001610
Code : NEW
Additional Details : Weight : 0.35000kg
 View more versions of this product
Product Code. 11817010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Alert:
To be disposed of when filters are saturated.
Description
Description
  • Product Type: Air-Purifying Respirator (APR)
  • Mask Type: Half Mask
  • Standards: EN405
  • Cartridge Type: Dual Cartridge, Pre-Assembled
  • Fastenings: One-Piece Sliding Suspension System
  • Hazards: Gases, Organic Vapors (Temp. >65°C), Inorganic Vapors, Acids, Ammoniac and Derived Aminates (Class 1); Solid and Liquid Particulates (Class 3)
  • For Use With (Equipment): Prefilter Pads (11847171, 12682743) and Pad Retaining Covers (12662653)
  • For Use With (Applications): Chemical Industry, Construction Industry, Food Services, Manufacturing Industry, Medical Industry, Pharmaceutical Industry
Specifications
Specifications
A1B1E1K1P3
EC 394, CEN TC79, EN 405
Cup/Molded
Respiratory Protection, Respiratory Protection
Prefilter Pads [11847171 (P1), 12682743 (P2)] and Pad Retaining Covers (12662653)
One-Piece Sliding Suspension System
EC 394, CEN TC79 EN 405, EC 394, CEN TC79 EN 405
Green, Yellow, Gray, Brown, Green, Yellow, Gray, Brown
Reusable
For protection against gas and organic vapors, For protection against gas and organic vapors
1 Ea., 1 Ea.