missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Mont-Blanc S3 Safety Shoes

Product Code. 11831771
Click to view available options
Size:
37
38
39
40
41
42
44
45
46
47
Unit Size:
Each
Pack of 2
This item is not returnable. View return policy

Product Code. 11831771

Brand: Honeywell™ 623321540/7

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance European Directive 89.686 CEE, EN 345-1, ISO 9001:2008
Color White
For Use With (Application) For food services, pharmaceutical and medical use
Material Leather
Size 40
Sole Features Anti-puncture Steel midsole, white color, made up of double density Polyurethane
Sole Material Double density Polyurethane
Style Slip-on safety
Toe Material Metallic
Resistant To Impact, splash, slip-resistant
Product Type Safety Shoe
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.