missing translation for 'onlineSavingsMsg'
Learn More

Sperian™ Honeywell™ Mont Blanc Safety Shoes

Product Code. 12681293
Change view
Click to view available options
Size:
35
36
39
41
42
44
45
46
47
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12681293 36 Each
12641303 42 Each
12611303 39 Each
12661303 44 Each
12671303 45 Each
12631303 41 Each
12671293 35 Each
12681303 46 Each
12691303 47 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 12681293 Supplier Sperian™ Supplier No. 623321636/7

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Moccasin-style slip-on leather shoe with limited stitching on the upper. Sperian™ Honeywell™ Mont Blanc Safety Shoes have a double density injected outsole for extra protection.

  • Lined and padded collar
  • Anti-puncture midsole
  • Metallic toe cap 200J
  • Perforated insole, polyurethane white
  • Protects against impacts, slips and splashes
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance European Directive 89.686 CEE, EN 345-1, ISO 9001:2008
Color White
For Use With (Application) For food services, pharmaceutical and medical use
Material Leather
Size 36
Sole Features Anti-puncture Steel midsole, white color, made up of double density Polyurethane
Sole Material Double density Polyurethane
Style Slip-on safety
Toe Material Metallic
Resistant To Impact, splash, slip-resistant
Product Type Safety Shoe
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.