Learn More
Honeywell KCL™ Honeywell KCL™ Sahara™ Plus 101 Gloves
Description
- Glove surface free from silicone (lacquer indifference, test method automotive industry)
- Leading glass manufacturer confirms surface compatibility for glass processing
Specifications
Specifications
| Certifications/Compliance | 971089 |
| Color | Yellow |
| Size | 9 |
| Thickness (Metric) | 0.8 cm |
| Length (Metric) | 240-270 cm |