missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell KCL™ Honeywell KCL™ 740-ermatril™ Gloves
Description
- AQL<0.65 free from natural latex resistant against a multitude of cytostatic drugs
- Virus-resistant according to ASTM 1671:2007
- Disposable protective glove excellent sense of touch
- Extremely comfortable
- Increase tear resistance
- Roughened fingertips for improved grip
Specifications
Specifications
| Certifications/Compliance | IFA 981069 |
| Description | Single-use powder-free nitrile glove |
| Disposable | Single-use |
| Latex Free | Yes |
| Material | Nitrile |
| Powdered | Powder Free |
| Product Type | Gloves |
| Size | 7 |
| Thickness (Metric) | 0.11 cm |
| Glove Cuff Color | Blue |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?