missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Honeywell™ Electrosoft Isoarc Latex Gloves

Product Code. 12677603
Change view
Click to view available options
:
Size: 7
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12677603 Size: 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12677603 Supplier Honeywell Safety Products™ Supplier No. 209200207

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Protect against electrical and Arc risks. Honeywell™ Electrosoft Isoarc Latex Gloves are soft and thin for excellent dexterity and include a grip finish on the palm.

  • Insulating glove for electrical work, specifically for arc flash environment.
  • Dielectric natural latex composition on chlorinated interior for comfort.
  • External layer in durable and mechanically resistant orange polychloroprene.
  • Patterned finish for grip in damp conditions.
  • Anatomical shape for ease of flexing.
  • Chlorinated inside.
  • Category RC.
TRUSTED_SUSTAINABILITY

Specifications

Arc Flash ATPV 21.6 cal/cm2
Certifications/Compliance CE CATEGORY III Irreversible risks, EN 60903-2003, CEI 903-2002, IEC 61482-1-2, EC Category PPE 3, EC Certificate Number 7017, EC Attestation Number 7017, Iso Arc 7KV
Classification 0
Cleaning Procedure Use water and soap. Dry below 65°C with ambient humidity
Description Box test : 4kA/15 cm and 7kA/30 cm
Coating Material Polychloroprene
Color Orange
Exterior Finish Patterned
Material Latex
Product Type Gloves
Size 7
Thickness (Metric) 1.1 mm
For Use With (Application) Electrical industry
Length (Metric) 36 cm
Voltage Max: 1,000V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.