Learn More
Sperian™ Honeywell™ Aluminum Rd40 Filter
The range of Honeywell™ filters protects against a multitude of risks encountered in many environments. The aluminium filters with Rd40 threaded connectors are for use with full-face mask and/or half-mask depending on the selected reference.
Brand: Sperian™ 1786730
Code : NEW
Additional Details : Weight : 0.80000kg
Description
- A2B2E2K1 NO P3 Standard
- Protection against gases, vapours and particles
- Body made of aluminium provides optimum resistance
- RD40 connections in a wide range of head pieces
- Low breathing resistance
- Adapted design, clearing the field of vision
- Safety cover for storage of filters of up to six months
-
Specifications
Filter | |
Aluminum | |
Respiratory protection against organic gases and vapors with a boiling point below 65°C, inorganic gases and vapors, solid and liquid particles |
E.C. Declaration of Conformity - EC Category PPE-3, Quality Assurance - ISO 9001/2000, EC Certificate Number - 0070-003-079-06-98-0042, EC Attestation - EC Attestation, EC Attestation Number - 0070-003-079-06-98-0042 | |
Aluminum Filter | |
Mask |