missing translation for 'onlineSavingsMsg'
Learn More

Homovanillic Acid, TRC

Product Code. 17939994
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17939994 50 mg 50mg
17928104 1 g 1g
17997804 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17939994 Supplier Toronto Research Chemicals Supplier No. TRCH66950050MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 306-08-1
Molecular Formula C9 H10 O4
Molecular Weight (g/mol) 182.17
Synonym Benzeneacetic acid, 4-hydroxy-3-methoxy-, Acetic acid, (4-hydroxy-3-methoxyphenyl)- (6CI,8CI), 4-Hydroxy-3-methoxybenzeneacetic acid, (4-Hydroxy-3-methoxyphenyl)acetic acid, 3-Methoxy-4-hydroxyphenylacetic acid, 3'-Methoxy-4'-hydroxyphenylacetic acid, HVA, Homovanillic acid, NSC 16682, Vanilacetic acid
IUPAC Name 2-(4-hydroxy-3-methoxyphenyl)acetic acid
SMILES COc1cc(CC(=O)O)ccc1O

Specifications

InChI Formula InChI=1S/C9H10O4/c1-13-8-4-6(5-9(11)12)2-3-7(8)10/h2-4,10H,5H2,1H3,(H,11,12)
Formula Weight 182.0579
Percent Purity >95%
Quantity 50 mg
Chemical Name or Material Homovanillic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.