missing translation for 'onlineSavingsMsg'
Learn More

Homovanillic acid, 98%

Código de producto. 11408890
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Tamaño de la unidad:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
11408890 25 g 25g
11429663 1 g 1g
11439663 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11408890 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor A12441.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 306-08-1 | C9H10O4 | 182.175 g/mol

Fluorimetric reagent and major catecholamine metaboliteHomovanillic acid is used as a reagent in the determination of oxidative enzymes. It is associated with dopamine levels in the brain. Its presence supports a diagnosis of neuroblastoma and malignant pheochromocytoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water,dimethyl sulfoxide, ethanol, methanol and ethyl acetate.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 306-08-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00004350
InChI Key QRMZSPFSDQBLIX-UHFFFAOYSA-N
Synonym homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
PubChem CID 1738
ChEBI CHEBI:545959
IUPAC Name 2-(4-hydroxy-3-methoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)CC(=O)O)O

Especificaciones

Melting Point 140°C to 145°C
Quantity 25 g
Beilstein 2213447
Merck Index 14,4740
Solubility Information Soluble in water,dimethyl sulfoxide,ethanol,methanol and ethyl acetate.
Formula Weight 182.18
Percent Purity ≥98%
Chemical Name or Material Homovanillic acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.